1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 97%

Warning

Synonyms: 1-(((Benzyloxy)carbonyl)amino)cyclobutanecarboxylic acid

H62122-MD H62122-06 H62122-03
AAH62122-03EA 116.55 USD
AAH62122-03 AAH62122-06 AAH62122-MD
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 97%
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
Formula: C₁₃H₁₅NO₄
MW: 249.266 g/mol
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD03426163
CAS Number: 190004-53-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR