Methyl-3-oxocyclobutanecarboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-oxocyclobutanecarboxylate

H62110-06 H62110-MD H62110-03
AAH62110-06EA 484.99 USD
AAH62110-06 AAH62110-03 AAH62110-MD
Methyl-3-oxocyclobutanecarboxylate 97%
Methyl-3-oxocyclobutanecarboxylate
Formula: C₆H₈O₃
MW: 128.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00604222
CAS Number: 695-95-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR