3-(Hydroxymethyl)-5-methylisoxazole 97%

Warning

Synonyms: 5-Methyl-3-isoxazolemethanol3-Hydroxymethyl-5-methylisoxazole

H61948-03 H61948-MD
AAH61948-03EA 342.4 USD
AAH61948-03 AAH61948-MD
3-(Hydroxymethyl)-5-methylisoxazole 97%
3-(Hydroxymethyl)-5-methylisoxazole
Formula: C₅H₇NO₂
MW: 113.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085129
CAS Number: 35166-33-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR