N-Boc-DL-valine 98%

Synonyms: BOC-DL-Valine2-((tert-Butoxycarbonyl)amino)-3-methylbutanoic acid

H61908-06 H61908-14
AAH61908-14EA 214.91 USD
AAH61908-14 AAH61908-06
N-Boc-DL-valine 98%
N-Boc-DL-valine
Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00069973
CAS Number: 54895-12-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR