N-Ethyl-p-toluenesulfonamide 98%

Warning

Synonyms: N-Ethyl-4-methylbenzenesulfonamideN-Tosylethylaminep-Tolueneethylsulfonamide

H61905-22
AAH61905-22EA 64.4 USD
AAH61905-22
N-Ethyl-p-toluenesulfonamide 98%
N-Ethyl-p-toluenesulfonamide
Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
Boiling Pt: 208 °C/745 mm
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00048511
CAS Number: 80-39-7
EINECS: 201-275-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR