1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%

Warning

Synonyms: 1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

H61817-03 H61817-MD
AAH61817-03EA 77.36 USD
AAH61817-03 AAH61817-MD
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
Formula: C₈H₈N₂O₂S
MW: 196.224 g/mol
Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00126746
CAS Number: 25252-46-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR