Ethyl (4-fluorobenzoyl)acetate 95%

Synonyms: (4-Fluorobenzoyl)acetic Acid Ethyl EsterEthyl 3-(4-Fluorophenyl)-3-oxopropionate3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester

H61813-MD H61813-03 H61813-06
AAH61813-03EA 73.77 USD
AAH61813-03 AAH61813-06 AAH61813-MD
Ethyl (4-fluorobenzoyl)acetate 95%
Ethyl (4-fluorobenzoyl)acetate
Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
Melting Pt: 117…120 °C
Density: 1.174
Storage Temperature: Ambient
MDL Number: MFCD03093631
CAS Number: 1999-00-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR