5-(Methylthio)thiophene-2-carboxaldehyde 97%

Warning

Synonyms: 2-Formyl-5-(methylthio)thiophene

H61803-03 H61803-06
AAH61803-06EA 348.32 USD
AAH61803-06 AAH61803-03
5-(Methylthio)thiophene-2-carboxaldehyde 97%
5-(Methylthio)thiophene-2-carboxaldehyde
Formula: C₆H₆OS₂
MW: 158.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173744
CAS Number: 24445-35-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR