Isoquinoline-4-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Isoquinoline-4-carboxaldehyde

H61788-MD H61788-03
AAH61788-MDEA 36.1 USD
AAH61788-MD AAH61788-03
Isoquinoline-4-carbaldehyde 97%
Isoquinoline-4-carbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00829440
CAS Number: 22960-16-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR