3-Nitro-4-(trifluoromethoxy)aniline 95%

Warning

H61456-22 H61456-14
AAH61456-14EA 59.96 USD
AAH61456-14 AAH61456-22
3-Nitro-4-(trifluoromethoxy)aniline 95%
3-Nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃N₂O₃
Boiling Pt: 300 °C/760 mm
Density: 1.543
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD03840207
CAS Number: 2822-50-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR