5-Fluoro-2-methyl-1H-indene-3-acetic acid 97%

Danger

Synonyms: (5-Fluoro-2-methyl-1H-inden-3-yl)acetic acid

H61410-MD
AAH61410-MDEA 178.83 USD
AAH61410-MD
5-Fluoro-2-methyl-1H-indene-3-acetic acid 97%
5-Fluoro-2-methyl-1H-indene-3-acetic acid
Formula: C₁₂H₁₁FO₂
MW: 206.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01076194
CAS Number: 32004-66-3
EINECS: 250-891-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR