Imidazo[1,2-b]pyridazine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61377-06 H61377-14
AAH61377-06EA 47.69 USD
AAH61377-06 AAH61377-14
Imidazo[1,2-b]pyridazine 98%
Imidazo[1,2-b]pyridazine
Formula: C₆H₅N₃
MW: 119.12 g/mol
Melting Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD07782103
CAS Number: 766-55-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR