N-Boc-L-aspartic acid 4-cyclohexyl ester 95%

Synonyms: 4-Cyclohexyl N-Boc-L-aspartateN-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Cyclohexyl EsterN-Boc-L-aspartic Acid 4-Cyclohexyl Ester

H61240-22 H61240-06 H61240-14
AAH61240-06EA 36.87 USD
AAH61240-06 AAH61240-14 AAH61240-22
N-Boc-L-aspartic acid 4-cyclohexyl ester 95%
N-Boc-L-aspartic acid 4-cyclohexyl ester
Formula: C₁₅H₂₅NO₆
MW: 315.37 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00061996
CAS Number: 73821-95-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR