N-Boc-3-benzyloxymethyl-L-histidine 98%
Synonyms:
Boc-His(Bom)-OH
H61234-14
H61234-03
H61234-06
AAH61234-14EA
334.11
USD
AAH61234-14
AAH61234-03
AAH61234-06
N-Boc-3-benzyloxymethyl-L-histidine 98%
N-Boc-3-benzyloxymethyl-L-histidine
Formula:
C₁₉H₂₅N₃O₅ MW: 375.42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043133 CAS Number: 79950-65-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...