Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
H61158-03
H61158-06
AAH61158-03EA
177.99
USD
AAH61158-03
AAH61158-06
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 95%
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate
Formula:
C₈H₉ClN₂O₂ MW: 200.622 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425963 CAS Number: 188781-08-0 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...