Tenoxicam 97%

Danger

Synonyms: 4-hydroxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide

H60837-03 H60837-06 H60837-14
AAH60837-06EA 107.12 USD
AAH60837-06 AAH60837-03 AAH60837-14
Tenoxicam 97%
Tenoxicam
Formula: C₁₃H₁₁N₃O₄S₂
MW: 337.368 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083502
CAS Number: 59804-37-4
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR