4-Hydroxy-o-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Hydroxy-o-toluic acid
, 4-Hydroxy-2-methylbenzoic acid
H60810-09
H60810-09
H60810-03
AAH60810-03EA
33.25
USD
AAH60810-03
AAH60810-09
CAAAH60810-09
4-Hydroxy-o-toluic acid 98%
4-Hydroxy-o-toluic acid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 177…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD02182261 CAS Number: 578-39-2 EINECS: 209-422-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...