Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dicumyl Peroxide
, Bis(1-methyl-1-phenylethyl)peroxide
, Bis(alpha,alpha-dimethylbenzyl)peroxide
H60442-36
H60442-36
H60442-22
AAH60442-36EA
167.4
USD
AAH60442-36
CAAAH60442-36
AAH60442-22
Bis(α,α-dimethylbenzyl)peroxide (dicumyl peroxide) 98%
Bis(α,α-dimethylbenzyl) peroxide (Dicumyl peroxide)
Formula:
C₁₈H₂₂O₂ MW: 270.37 g/mol Melting Pt: 39…41 °C Density: 1.56 Storage Temperature: Ambient |
MDL Number:
MFCD00036227 EINECS: 201-279-3 UN: 3110 ADR: "5.2", |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...