1-Boc-4-(2-formylphenyl)piperazine 97%

Warning

Synonyms: 1-Boc-4-(2-Formylphenyl)piperazine

H55966-03 H55966-14 H55966-06
AAH55966-06EA 193.14 USD
AAH55966-06 AAH55966-03 AAH55966-14
1-Boc-4-(2-formylphenyl)piperazine 97%
1-Boc-4-(2-formylphenyl)piperazine
Formula: C16H22N2O3
MW: 290.36 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD05864664
CAS Number: 174855-57-3
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR