Tricyclohexyltin chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H55493-03 H55493-14 H55493-06
AAH55493-06EA 148.99 USD
AAH55493-06 AAH55493-03 AAH55493-14
Tricyclohexyltin chloride 97%
Tricyclohexyltin chloride
Formula: C₁₈H₃₃ClSn
MW: 403.62 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003817
CAS Number: 3091-32-5
EINECS: 221-437-5
UN: 3146
ADR: "6.1",III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR