Potassium-2-chlorophenyltrifluoroborate 95%

Warning

Synonyms: Potassium (2-chlorophenyl)(trifluoro)borate(1-)Borate(1-), (2-chlorophenyl)trifluoro-, potassium (1:1)

H37855-06 H37855-03
AAH37855-03EA 76.69 USD
AAH37855-03 AAH37855-06
Potassium-2-chlorophenyltrifluoroborate 95%
Potassium-2-chlorophenyltrifluoroborate
Formula: C₆H₄BClF₃K
MW: 218.46 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD09993172
CAS Number: 870195-98-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR