tert-Butyl-4-(bromomethyl)benzoate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
tert-Butyl 4-(bromomethyl)benzoate
, 4-Bromo-methyl-benzoic acid mono tert-butyl ester
H35540-03
H35540-09
AAH35540-03EA
79.79
USD
AAH35540-03
AAH35540-09
tert-Butyl-4-(bromomethyl)benzoate 95%
tert-Butyl 4-(bromomethyl)benzoate
Formula:
C₁₂H₁₅BrO₂ MW: 271.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973450 CAS Number: 108052-76-2 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...