2-(Carboxymethylthio)succinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-((Carboxymethyl)thio)succinic acid
A19818-18
A19818-09
AAA19818-09EA
26.19
USD
AAA19818-09
AAA19818-18
2-(Carboxymethylthio)succinic acid 97%
2-(Carboxymethylthio)succinic acid
Formula:
C₆H₈O₆S MW: 208.184 g/mol Melting Pt: 138…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002720 CAS Number: 99-68-3 EINECS: 202-778-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...