Antimony ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.500 in)

47181-20
AA47181-20EA 425.07 USD
AA47181-20
Antimony ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.500 in)
Antimony

Antimony rod, 12.7mm (0.500in) dia, 99.999% (metals basis), ^=2.16cm/20g, Cast rod.

Formula: Sb
MW: 121.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR