Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II) (14,4% Pd) ≥98%

Warning

Synonyms: [Bis(dicyclohexylphosphinophenyl)ether]dichloropalladium(II)[Bis(dicyclohexylphosphinophenyl)ether]palladium(II) dichloride

46872-03
AA46872-03EA 292.58 USD
AA46872-03
Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II) (14,4% Pd) ≥98%
Dichloro[bis(dicyclohexylphosphinophenyl)ether]palladium(II)
Formula: C₃₆H₅₂Cl₂OP₂Pd
MW: 740.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26967909
CAS Number: 1265140-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR