(Acetylacetonato)(norbornadiene)rhodium(I) Rh 34,9%
|
Warning
|
Synonyms:
(Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I)
46727-03
AA46727-03EA
437.59
USD
AA46727-03
(Acetylacetonato)(norbornadiene)rhodium(I) Rh 34,9%
(Acetylacetonato)(norbornadiene)rhodium(I)
Formula:
C₁₂H₁₅O₂Rh MW: 294.16 g/mol Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075071 CAS Number: 32354-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...