Pharmabotics Laparoscopic Trainer
Supplier: Pharmabiotics
1005141 1005139 1005140 1005143 1005142
470119-542EA 245 USD
470119-542 470119-538 470119-540 470119-546 470119-544
Pharmabotics Laparoscopic Trainer
Models Human Biology Models Human Reproductive System Models
Portable Laparoscopic Training
  • Easy to store size
  • Multiple skin options
The Portable Laparoscopic Trainer is designed to provide low cost assistance in laparoscopic training. Supplied with 4mm black Neoprene skin. Simple to assemble and ready for use in seconds. Works with wet tissue (e.g. Pig liver) or dry (e.g. foam).
Ordering information: This kit includes 4 mm black Neoprene skin.