Pharmabotics Skin Suture Training
1005134 1005133
470119-568EA 334 USD
470119-568 470119-566
Pharmabotics Skin Suture Training
Models Human Biology Models
Basic Skin Suturing
  • Replaceable pad
  • Sub dermal layer allows for intra-dermal suturing
  • Secure mounting onto smooth surfaces
A great low cost way to practice surgical opening and closing techniques. Features . Economical reversible pad. The material has been designed to make complex incisions such as "flap and dog turn" suturing more realistic . A sub-dermal layer is present so that it is possible to make secure intra-dermal sutures . Base has suction feet to enable slip free use.