Female Pelvic Contraceptive Model
1017935
470120-392EA 194.75 USD
470120-392
Female Pelvic Contraceptive Model
Models Human Biology Models
Compact, Economical Training Tool
  • Demonstrate and practice most common female contraceptives
  • Side viewing port allows for accurate assessment of cervical coverage
This female contraceptive model, made from soft BIOlike™ material, provides a  cross-section of a female pelvis for demonstrating proper contraceptive insertion. The side cutout lets viewers see and understand contraceptive placement and removal from the female genitals. Great tool for sex education!
Ordering information: Contraceptives not included.