Anti-RAB3GAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24599-1-AP-150UL
10093-202EA 402 USD
10093-202
Anti-RAB3GAP2 Rabbit Polyclonal Antibody
Antibodies
RAB3GAP2, also known as RAB3-GAP150, belongs to the Rab3-GAP regulatory subunit family. Rab3 GAP consists of two subunits: the catalytic subunit p130 and the noncatalytic subunit p150. Rab3 GAP is ubiquitously expressed and enriched in the synaptic soluble fraction of brain, consistent with it having a key role in neurodevelopment. RAB3-GAP150 forms the Rab3 GTPase-activating complex with RAB3-GAP130,where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. Defects in RAB3-GAP150 are the cause of Martsolf and Warburg Micro syndrome.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: RAB3GAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR