Anti-LCTL Rabbit Polyclonal Antibody

Supplier: Proteintech
24589-1-AP-150UL
10089-562EA 402 USD
10089-562
Anti-LCTL Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF-7 Cells, 1:500-1:5000; IF: HeLa Cells, 1:10-1:100

Type: Primary
Antigen: LCTL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR