Anti-SLC37A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24582-1-AP-150UL
10094-768EA 402 USD
10094-768
Anti-SLC37A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF-7 Cells, 1:200-1:2000

Type: Primary
Antigen: SLC37A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR