Anti-HEATR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
24578-1-AP-150UL
10088-138EA 402 USD
10088-138
Anti-HEATR2 Rabbit Polyclonal Antibody
Antibodies
HEATR2 also termed as HEAT repeat containing 2 is an 855 amino acid protein. HEATR2 is a member of a family of ten other uncharacterized HEAT-repeat-containing proteins, which contains HEAT (Huntingtin, elongation factor 3 (EF3), protein phosphatase 2A (PP2A) and the yeast PI3-kinase Tor1) repeats. HEAT repeats form rod-like helical structures that are involved in intracellular transport. HEATR2 play an important role in dynein arm transportation or assembly. HEATR2 mutation is implicated in Primary ciliary dyskinesia 18.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human lung Tissue, 1:20-1:200; IF: HEK-293 Cells, 1:10-1:100

Type: Primary
Antigen: HEATR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR