Anti-CD41/Integrin alpha 2b Rabbit Polyclonal Antibody

Supplier: Proteintech
24552-1-AP-150UL
10088-996EA 402 USD
10088-996
Anti-CD41/Integrin alpha 2b Rabbit Polyclonal Antibody
Antibodies
The integrins are a superfamily of cell adhesion receptors that bind to extracellular matrix ligands, cell-surface ligands, and soluble ligands . They are transmembrane αβ heterodimers and at least 24 distinct integrin heterodimers are formed by the combination of 18 α and eight β known subunits . In addition to mediating cell adhesion, integrins also play important roles in modulating signal transduction pathways that control cellular responses including migration, proliferation, differentiation, and apoptosis . Integrin alpha-IIb (ITGA2B, CD41) is expressed on platelets, megakaryocytes and some hematopoietic progenitor cells . This protein undergoes post-translational cleavage to yield disulfide linked light and heavy chains which join with integrin beta3 (CD61) to form a receptor for fibronectin, fibrinogen, plasminogen, prothrombin, thrombospondin and vitronectin. Integrin alpha-IIb/beta-3 is crucial for platelet aggregation through binding of soluble fibrinogen.

Western Blot: Human Blood Tissue, 1:1000-1:10000; IHC: Human spleen, 1:20-1:200

Type: Primary
Antigen: CD41/Integrin alpha 2b
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR