Anti-TBCEL Rabbit Polyclonal Antibody

Supplier: Proteintech
24548-1-AP-150UL
10095-644EA 402 USD
10095-644
Anti-TBCEL Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:1000-1:10000

Type: Primary
Antigen: TBCEL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR