Anti-PTPN6 Rabbit Polyclonal Antibody

Supplier: Proteintech
24546-1-AP-150UL
10094-630EA 402 USD
10094-630
Anti-PTPN6 Rabbit Polyclonal Antibody
Antibodies
PTPN6(Tyrosine-protein phosphatase non-receptor type 6) is also named as HCP, PTP1C and belongs to the protein-tyrosine phosphatase family. It regulates muscle insulin action in a cell-autonomous manner, further suggesting that the PTPase negatively modulates insulin action through down-regulation of both insulin signaling to AKT1 and SLC2A4 translocation, as well as SLC2A4 expression. It has 4 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: PTPN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR