Anti-ZNF140 Rabbit Polyclonal Antibody

Supplier: Proteintech
24531-1-AP-150UL
10097-302EA 402 USD
10097-302
Anti-ZNF140 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SKOV-3 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ZNF140
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR