Anti-C16ORF72 Rabbit Polyclonal Antibody

Supplier: Proteintech
24522-1-AP-150UL
10083-784EA 402 USD
10083-784
Anti-C16ORF72 Rabbit Polyclonal Antibody
Antibodies
C16orf72 is identified on chromosome 16p13.2 loci. The MW of C16orf72 is 25-35 kDa in our detection.

Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: C16ORF72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR