Anti-CPS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24513-1-AP-150UL
10082-052EA 402 USD
10082-052
Anti-CPS1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: CPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR