Anti-ZNF512 Rabbit Polyclonal Antibody

Supplier: Proteintech
24508-1-AP-150UL
10097-356EA 402 USD
10097-356
Anti-ZNF512 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse small intestine Tissue, 1:200-1:2000

Type: Primary
Antigen: ZNF512
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR