Anti-RRP7A Rabbit Polyclonal Antibody

Supplier: Proteintech
24490-1-AP-150UL
10094-008EA 402 USD
10094-008
Anti-RRP7A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Thymus Tissue, 1:500-1:5000; IF:A431 Cells, 1:10-1:100; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: RRP7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR