Anti-AMZ1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24484-1-AP-150UL
10082-872EA 402 USD
10082-872
Anti-AMZ1 Rabbit Polyclonal Antibody
Antibodies
AMZ1, also named as KIAA1950, is a new human Zinc metalloproteases. It exhibits aminopeptidase activity against neurogranin in vitro. AMZ1 does not hydrolyze angiotensin-2. It is mainly detected in liver and heart. AMZ1 has some isoforms with MW 55 kDa, 33 kDa, 30 kDa and 28 kDa.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: AMZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR