Anti-EXOSC9 Rabbit Polyclonal Antibody

Supplier: Proteintech
24470-1-AP-150UL
10086-728EA 524.48 USD
10086-728
Anti-EXOSC9 Rabbit Polyclonal Antibody
Antibodies
EXOSC9, also named as PMSCL1 and PM/Scl-75, is a key subunit of the exosome complex. EXOSC9 maintains self-renewal in a cell-autonomous manner. EXOSC9 has 4 isoforms with MW 49 kDa, 51 kDa, 39 kDa and 41 kDa. It’s polymyositis/scleroderma auto-antigen 75 kDa. The MW of EXOSC9 is migrated to 60-75 kDa for modification in WB detection.

IHC: Human prostate hyperplasia Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100; Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: EXOSC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR