Anti-CEP44 Rabbit Polyclonal Antibody

Supplier: Proteintech
24457-1-AP-150UL
10084-652EA 524.48 USD
10084-652
Anti-CEP44 Rabbit Polyclonal Antibody
Antibodies
CEP44, also named as KIAA1712 and PS1TP3, localizes to centrosomes, spindle poles, and weakly in midbody. It is a 44 kDa centrosome protein.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: CEP44
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR