Anti-NUP205 Rabbit Polyclonal Antibody

Supplier: Proteintech
24439-1-AP-150UL
10091-410EA 402 USD
10091-410
Anti-NUP205 Rabbit Polyclonal Antibody
Antibodies
NUP205, also named as C7orf14 and KIAA0225, is a 205 kDa nucleoporin. It plays a role in the nuclear pore complex (NPC) assembly and/or maintenance.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: NUP205
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR