Anti-BRIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
24436-1-AP-150UL
10081-984EA 402 USD
10081-984
Anti-BRIP1 Rabbit Polyclonal Antibody
Antibodies
BRIP1 also named as BACH1, FANCJ or ATP dependent RNA helicase BRIP1 is a 1249 amino acid protein, which contains one helicase ATP-binding domain and belongs to the DEAD box heilicase family, DEAH subfamily. BRIP1 contains nuclear localization signal and is ubiquitously expressed with highest levels in testis, an expression pattern similar to that of BRCA1. BRIP1 is required to resolve the secondary structures of guanine-rich DNA that occasionally form during lagging-strand DNA synthesis.

Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: BRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR