Anti-LYAR Rabbit Polyclonal Antibody

Supplier: Proteintech
24433-1-AP-150UL
10089-428EA 402 USD
10089-428
Anti-LYAR Rabbit Polyclonal Antibody
Antibodies
LYAR also named as ZLYAR or Cell growth regulating nucleolar protein is 379 amino acid protein, which contains 2 C2H2-type zinc fingers. LYAR is expressed at at high levels in immature spermatocytes, early embryos and in fetal liver and thymus ,with low expression in kidney and spleen, LYAR may function as a novel nucleolar oncoprotein to regulate cell growth. It is suggested that LYAR participates in regulating the stability of C23, a protein that is critical for maintaining the self-renewal and differentiation of embryonic stem cells.

Western Blot:A431 Cells, 1:500-1:5000

Type: Primary
Antigen: LYAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR