Anti-TEX38 Rabbit Polyclonal Antibody

Supplier: Proteintech
24429-1-AP-150UL
10095-810EA 402 USD
10095-810
Anti-TEX38 Rabbit Polyclonal Antibody
Antibodies
TEX38, also named as ATPAF1-AS1 and C1orf223, is ATPAF1 antisense gene protein 1. This antibody detects 25-28 kDa with mouse testis and MDA-MB-453s cell lysate.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: TEX38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR