Anti-C10ORF27 Rabbit Polyclonal Antibody

Supplier: Proteintech
24424-1-AP-150UL
10083-744EA 402 USD
10083-744
Anti-C10ORF27 Rabbit Polyclonal Antibody
Antibodies
C10orf27, also named as TBATA and SPATIAL, may play a role in spermatid differentiation. We always got 40-45 kDa band in our detection.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: C10ORF27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR