Anti-TALPID3 Rabbit Polyclonal Antibody

Supplier: Proteintech
24421-1-AP-150UL
10095-600EA 402 USD
10095-600
Anti-TALPID3 Rabbit Polyclonal Antibody
Antibodies
KIAA0586 also named as Talpid3 is a 1472 amino-acid protein, which belongs to Talpid3 family. Talpid3 is a gene essential for vertebrate development. Studies in chicken, mouse, zebrafish and human have shown that TALPID3 is a centrosomal protein, mutants for TALPID3 fail to produce cilia, resulting in far reaching signalling defects such as polydactyly, dorsalised neural tube, craniofacial and vascular defects often due to abnormalities in Hedgehog signal transduction. Talpid3 is absolutely required for the function of both Gli repressor and activator in the intracellular Hedgehog pathway and ciliogenesis. TALPID3 may be identified as a subcellular maker of centrosome and needed to further studied.

Western Blot: Mouse Embryo Tissue, 1:200-1:2000

Type: Primary
Antigen: TALPID3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR